missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane, 94%

Product Code. 11312076
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11312076

Brand: Thermo Scientific Alfa Aesar L03239.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1719-53-5 | C4H10Cl2Si | 157.11 g/mol

Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water and ether.

Notes
Moisture Sensitive. Store at 4°C. Protect from moisture.Incompatible materials are Oxidizing agents, Bases, Water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-53-5
Molecular Formula C4H10Cl2Si
Molecular Weight (g/mol) 157.11
MDL Number MFCD00000493
InChI Key BYLOHCRAPOSXLY-UHFFFAOYSA-N
Synonym diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
PubChem CID 15589
IUPAC Name dichloro(diethyl)silane
SMILES CC[Si](Cl)(Cl)CC

Specifications

Boiling Point 129°C to 131°C
Melting Point -97°C
Quantity 50 g
UN Number UN1767
Beilstein 605313
Flash Point 28°C (82°F)
Solubility Information Slightly soluble in water and ether.
Formula Weight 157.12
Percent Purity 94%
Odor Acrid
Refractive Index 1.431
Sensitivity Moisture sensitive
Density 1.055
Chemical Name or Material Dichlorodiethylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.