missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(triphenylphosphine) Palladium (II), TRC

Product Code. 30384923
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384923 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384923 Supplier Toronto Research Chemicals Supplier No. TRCD4341451G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Quantity 1 g
Synonym Bis(triphenylphosphine)dichloropalladium, Bis(triphenylphosphine)palladium Chloride, Bis(triphenylphosphine)palladium Dichloride, Bis(triphenylphosphine)palladium(II) Chloride, Bis(triphenylphosphine)palladium(II) Dichloride, Bis(triphenylphosphino)palladium Chloride, Dichlorobis(triphenylphosphine)palladium, Dichlorobis(triphenylphosphine)palladium(II), NSC 122924, Palladiumbis(triphenylphosphine) Dichloride, PdCl2(PPh3)2
InChI Formula InChI=1S/2C18H15P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2
SMILES [Cl-][Pd+2]([Cl-])([P](C=1C=CC=CC1)(C=2C=CC=CC2)C=3C=CC=CC3)[P](C=4C=CC=CC4)(C=5C=CC=CC5)C=6C=CC=CC6
IUPAC Name dichloropalladium;triphenylphosphane
Molecular Weight (g/mol) 701.89
Formula Weight 700.0235
Chemical Name or Material Dichlorobis(triphenylphosphine) Palladium (II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.