missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(triphenylphosphine)nickel(II), 98%

Product Code. 11341469
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341469 10 g 10g
11351469 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341469 Supplier Thermo Scientific Alfa Aesar Supplier No. 013930.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol

Dichlorobis(triphenylphosphine)nickel(II) is used as a catalyst for cross-coupling of Grignard reagents, hydrosilylations, hydrogenation and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14264-16-5
Molecular Formula C36H30Cl2NiP2
Molecular Weight (g/mol) 654.18
MDL Number MFCD00009592 MFCD00003043 MFCD20489348
InChI Key VHQPRJDILRYZBG-UHFFFAOYSA-L
Synonym bis triphenylphosphine dichloronickel ii
PubChem CID 131664342
IUPAC Name nickel(2+);triphenylphosphane;dihydrochloride
SMILES Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼225°C (decomposition)
Color Black to Gray
Odor Odorless
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 654.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Dichlorobis(triphenylphosphine)nickel(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.