missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorobis(triphenylphosphine)nickel(II), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Dichlorobis(triphenylphosphine)nickel(II) is used as a catalyst for cross-coupling of Grignard reagents, hydrosilylations, hydrogenation and polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
Specifications
Melting Point | ∼225°C (decomposition) |
Color | Black to Gray |
Odor | Odorless |
Quantity | 10 g |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 654.18 |
Percent Purity | 98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Dichlorobis(triphenylphosphine)nickel(II) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'