missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(ethylenediamine)platinum(II), Pt 50%

Product Code. 11470221
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470221 0.25 g 0.25g
11333688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470221 Supplier Thermo Scientific Alfa Aesar Supplier No. 039280.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21430-85-3 | C4H16Cl2N4Pt | 386.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21430-85-3
Molecular Formula C4H16Cl2N4Pt
Molecular Weight (g/mol) 386.18
MDL Number MFCD00009881
InChI Key MXLIXEZAMQLDMM-UHFFFAOYSA-L
Synonym Bis(ethylenediamine)platinum(II) chloride
IUPAC Name platinum(2+) bis(ethane-1,2-diamine) dichloride
SMILES [Cl-].[Cl-].[Pt++].NCCN.NCCN

Specifications

Melting Point 300°C (decomposition)
Color White
Linear Formula PtCl2(H2NCH2CH2NH2)2
Quantity 0.25 g
Formula Weight 386.18
Concentration or Composition (by Analyte or Components) Pt 50%
Physical Form Crystalline
Chemical Name or Material Dichlorobis(ethylenediamine)platinum(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.