missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 16.1%

Product Code. 15476925
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476925 1 g 1g
15486925 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476925 Supplier Thermo Scientific Alfa Aesar Supplier No. 045453.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34409-44-4 | C28H46Cl2P2Pd | 621.94 g/mol

Dichlorobis(di-tert-butylphenylphosphine)palladium(II) is used as an effective catalyst for cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34409-44-4
Molecular Formula C28H46Cl2P2Pd
Molecular Weight (g/mol) 621.94
MDL Number MFCD09953450
InChI Key DFKFLLBQLYGVIH-UHFFFAOYSA-L
Synonym dichlorobis di-tert-butylphenylphosphine palladium ii, bis di-t-butyl phenyl phosphine palladium dichloride, bis di-tert-butyl phenyl phosphane ; palladium chloride
PubChem CID 56649816
IUPAC Name ditert-butyl(phenyl)phosphane;dichloropalladium
SMILES Cl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C

Specifications

Melting Point (decomposition)
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water and polar organic solvents.
Formula Weight 621.95
Physical Form Solid
Chemical Name or Material Dichlorobis(di-tert-butylphenylphosphine)palladium(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.