missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II), 97%, Thermo Scientific™

Product Code. 10331140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10331140 250 mg 250mg
10491810 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10331140 Supplier Thermo Scientific Acros Supplier No. 435272500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 911128-25-1
Molecular Formula C30H36Cl2N2P2Ru
Molecular Weight (g/mol) 658.55
MDL Number MFCD15144861
InChI Key MLZFSEGMJUGESY-UHFFFAOYSA-L
PubChem CID 71310857
IUPAC Name dichlororuthenium;3-diphenylphosphanylpropan-1-amine
SMILES C1=CC=C(C=C1)P(CCCN)C2=CC=CC=C2.C1=CC=C(C=C1)P(CCCN)C2=CC=CC=C2.Cl[Ru]Cl

Specifications

Color Yellow to Brown
Elemental Analysis 52.5 to 56.9% C, 4.1 to 4.4% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 658.55
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
compliance-icons
Product Identifier
  • Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.