missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetyl chloride, 97%

Product Code. p-7026341
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11495966

Brand: Thermo Scientific Alfa Aesar B22296.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-36-7 | C2HCl3O | 147.379 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-36-7
Molecular Formula C2HCl3O
Molecular Weight (g/mol) 147.379
MDL Number MFCD00000840
InChI Key FBCCMZVIWNDFMO-UHFFFAOYSA-N
Synonym dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
PubChem CID 6593
ChEBI CHEBI:34688
IUPAC Name 2,2-dichloroacetyl chloride
SMILES C(C(=O)Cl)(Cl)Cl

Specifications

Density 1.532
Boiling Point 107°C to 108°C
Flash Point None
Refractive Index 1.462
Quantity 100 g
UN Number UN1765
Sensitivity Moisture sensitive
Merck Index 14,3053
Formula Weight 147.39
Percent Purity 97%
Chemical Name or Material Dichloroacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.