missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetonitrile, TRC

Product Code. 30370305
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370305 100 g
30378841 10 g
30378682 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370305 Supplier Toronto Research Chemicals Supplier No. TRCD431740100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3018-12-0
Molecular Formula C2 H Cl2 N
Molecular Weight (g/mol) 109.94
Synonym Dichloroacetonitrile, Dichloromethyl cyanide, NSC 60511
IUPAC Name 2,2-dichloroacetonitrile
SMILES ClC(Cl)C#N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C2HCl2N/c3-2(4)1-5/h2H
Formula Weight 108.9486
Chemical Name or Material Dichloroacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.