missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog Number 11463304
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3018-12-0
C2HCl2N
109.937
MFCD00040886
STZZWJCGRKXEFF-UHFFFAOYSA-N
dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562
18177
CHEBI:82444
2,2-dichloroacetonitrile
C(#N)C(Cl)Cl
This item is not returnable. View return policy
3018-12-0
C2HCl2N
109.937
MFCD00040886
STZZWJCGRKXEFF-UHFFFAOYSA-N
dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562
18177
CHEBI:82444
2,2-dichloroacetonitrile
C(#N)C(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.369
Boiling Point 112°C to 114°C
Flash Point 35°C (95°F)
Odor Pungent
Linear Formula Cl2CHCN
Refractive Index 1.4405
Quantity 5 g
UN Number UN2920
Beilstein 1739029
Formula Weight 109.94
Percent Purity ≥98%
Chemical Name or Material Dichloroacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'