missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic Acid, TRC

Product Code. 30136227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136227 5 g 5g
30134736 1 g 1g
18093730 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136227 Supplier Toronto Research Chemicals Supplier No. TRCD4318255G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2 H2 Cl2 O2
Molecular Weight (g/mol) 128.94
Synonym Acetic acid, dichloro- (5CI,8CI,9CI), 2,2-Dichloroacetic acid, DCA, DCA (acid), DCAA, DKhUK, Dichloracetic acid, Dichlorethanoic acid, Dichloroacetic acid, Dichloroethanoic acid, NSC 2654
IUPAC Name 2,2-dichloroacetic acid
SMILES OC(=O)C(Cl)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)
Formula Weight 127.9432
Percent Purity >95%
Chemical Name or Material Dichloroacetic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.