missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic Acid-d2, TRC

Product Code. 30383527
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383527 50 mg 50mg
30380717 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30383527 Supplier Toronto Research Chemicals Supplier No. TRCD43182650MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloroacetic Acid-d2
CAS 63403-57-6
Purity Grade Notes GC
Molecular Formula C2 D2 Cl2 O2
Quantity 50 mg
Synonym Acetic-d acid-d, dichloro- (9CI), 2,2-Dichloroacetic-2-d acid-d, Dichloroacetic acid-d2, Dichloroacetic Acid-D2
InChI Formula InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i1D/hD
SMILES [2H]OC(=O)C([2H])(Cl)Cl
IUPAC Name deuterio 2,2-dichloro-2-deuterioacetate
Molecular Weight (g/mol) 130.9544
Formula Weight 129.9557
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.