missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic acid, 99%, Thermo Scientific Chemicals

Product Code. 11457786
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457786 2500 g 2500g
11447786 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457786 Supplier Thermo Scientific Alfa Aesar Supplier No. A14740.0E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-43-6 | C2H2Cl2O2 | 128.936 g/mol

Dichloroacetic acid salts are used as drugs due to its inhibition of the enzyme pyruvate dehydrogenase kinase. It is also employed to stem the production of lactic acid to treat lactic acidosis. It is clinically essential in children to treat lactic acidosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ethanol, diethyl ether, n-octanol and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
Synonym dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
PubChem CID 6597
ChEBI CHEBI:36386
IUPAC Name 2,2-dichloroacetic acid
SMILES C(C(=O)O)(Cl)Cl

Specifications

Melting Point 9°C to 11°C
Density 1.564
Boiling Point 192°C to 194°C
Flash Point >110°C (230°F)
Assay Percent Range 99%
Odor Pungent
Refractive Index 1.462
Quantity 2500 g
UN Number UN1764
Beilstein 1098596
Sensitivity Hygroscopic
Merck Index 14,3050
Solubility Information Miscible with water,ethanol,diethyl ether,n-octanol and acetone.
Formula Weight 128.94
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dichloroacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.