missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%

Product Code. 11397768
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397768 1 g 1g
11307778 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397768 Supplier Thermo Scientific Alfa Aesar Supplier No. 033657.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12354-85-7 | C20H30Cl4Rh2 | 618.07 g/mol

Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer is used as a catalyst for amidation, reductive alkylation, hydrogenation, chiral hydrogenation reactions. It is also used as a catalyst for oxidative olefination reactions, C-C bond cleavage of secondary alcohols, oxidative annulation of pyridine and ortho C-H olefination of phenol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12354-85-7
Molecular Formula C20H30Cl4Rh2
Molecular Weight (g/mol) 618.07
MDL Number MFCD00061552
InChI Key QNIVKTTWBMFSBR-UHFFFAOYSA-J
Synonym Pentamethylcyclopentadienylrhodium(III) chloride dimer
PubChem CID 74222901
IUPAC Name 1,2,3,4,5-pentamethylcyclopenta-1,3-diene;rhodium(2+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Rh++].[Rh++].C[C]1C(C)=C(C)C(C)=C1C.C[C]1C(C)=C(C)C(C)=C1C

Specifications

Melting Point >300°C
Color Red
Odor Odorless
Quantity 1 g
Solubility Information Soluble in chloroform and acetone. Slightly soluble in tetrahydrofuran and methanol. Insoluble in water and diethylether
Formula Weight 618.08
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.