Learn More
Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, Thermo Scientific Chemicals
POA
Chemical Identifiers
CAS | 12354-84-6 |
---|---|
Molecular Formula | C20H30Cl4Ir2 |
Molecular Weight (g/mol) | 796.694 |
MDL Number | MFCD00075435 |
InChI Key | MMAGMBCAIFVRGJ-UHFFFAOYSA-J |
Synonym | unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di |
PubChem CID | 76030743 |
IUPAC Name | iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride |
SMILES | C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15457005
|
Thermo Scientific Alfa Aesar
045565.ME |
500 mg |
POA
500mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15447005
|
Thermo Scientific Alfa Aesar
045565.04 |
2 g |
POA
2g |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer is used as a precursor to catalysts for the asymmetric transfer hydrogenation of ketones. It is a catalyst for greener amine synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12354-84-6 | |
796.694 | |
MMAGMBCAIFVRGJ-UHFFFAOYSA-J | |
76030743 | |
C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3] |
C20H30Cl4Ir2 | |
MFCD00075435 | |
unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di | |
iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride |
Specifications
12354-84-6 | |
MFCD00075435 | |
MMAGMBCAIFVRGJ-UHFFFAOYSA-J | |
iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride | |
76030743 | |
Dichloro (pentamethylcyclopentadienyl)iridium (III) dimer |
C20H30Cl4Ir2 | |
unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di | |
C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3] | |
796.694 | |
796.67 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only