missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(ethylenediamine)palladium(II), Pd 44.8%

Product Code. 11325998
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325998 5 g 5g
11335998 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325998 Supplier Thermo Scientific Alfa Aesar Supplier No. 010496.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15020-99-2 | C2H8Cl2N2Pd | 237.42 g/mol

Catalyst for C-C coupling reactions like Sonogashira coupling. Formation of host-guest complexes. Synthesis of mixed-metal, mixed-pyrimidine self-assembling metallacalix[n]arenes; ring-in-ring complex; binding of metal with glycose phosphates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15020-99-2
Molecular Formula C2H8Cl2N2Pd
Molecular Weight (g/mol) 237.42
MDL Number MFCD00044991
InChI Key CAYKJANQVKIYPJ-UHFFFAOYSA-L
Synonym dichloro 1,2-diaminoethane palladium, dichloro ethylenediamine palladium ii, dichloro 1,2-diaminoethane ;palladium, dichloro ethylenediamine palladate ii, ethylenediamine palladium ii chloride, ethylenediamine; palladium chloride, palladium ii chloride ethylenediamine complex, dichloro ethylenediamine palladium ii , pd
PubChem CID 11139157
SMILES Cl[Pd]Cl.NCCN

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Very low solubility in water.
IUPAC Name dichloropalladium;ethane-1,2-diamine
Formula Weight 237.41
Concentration or Composition (by Analyte or Components) Pd 44.8%
Physical Form Crystalline
Chemical Name or Material Dichloro(ethylenediamine)palladium(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.