missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(diethylamino)phosphine, 97%

Product Code. 11495690
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11495690

Brand: Thermo Scientific Alfa Aesar H25934.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1069-08-5 | C4H10Cl2NP | 174.005 g/mol

Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.

Solubility
Reacts violently in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-08-5
Molecular Formula C4H10Cl2NP
Molecular Weight (g/mol) 174.005
MDL Number MFCD00013614
InChI Key BPEMCEULJQTJMI-UHFFFAOYSA-N
Synonym diethylphosphoramidous dichloride, dichloro diethylamino phosphine, diethylaminodichlorophosphine, et2npcl2, acmc-2098mq, diethylamido dichlorophosphite, diethylamino dichlorophosphine, dichloro diethylamido phosphine, dichlorophosphanyl diethylamine, dichlorophosphorous diethylamide
PubChem CID 2733289
IUPAC Name N-dichlorophosphanyl-N-ethylethanamine
SMILES CCN(CC)P(Cl)Cl

Specifications

Density 1.196
Boiling Point 179°C
Flash Point 83°C (181°F)
Odor Irritating
Linear Formula (CH3CH2)2NPCl2
Refractive Index 1.497
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts violently in water.
Formula Weight 174.01
Percent Purity 97%
Chemical Name or Material Dichloro(diethylamino)phosphine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.