missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(cycloocta-1,5-diene)ruthenium(II) Polymer, TRC

Product Code. 30381705
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381705 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381705 Supplier Toronto Research Chemicals Supplier No. TRCD43654510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloro(cycloocta-1,5-diene)ruthenium(II) Polymer
CAS 50982-12-2
Molecular Formula [C8H10Cl2Ru ].x
Quantity 10 g
Synonym Dichloro(1,5-cyclooctadiene)-ruthenium, 1,5-Cyclooctadiene, Ruthenium Complex, (1,5-Cyclooctadiene)ruthenium(II) Chloride Polymer, 1,5-Cyclooctadienedichlororuthenium, Cyclooctadieneruthenium Dichloride, Dichloro(1,5-cyclooctadiene)ruthenium, Dichloro(cyclooctadiene)ruthenium, Dichloro[(1,2,5,6-eta)-1,5-cyclooctadiene]ruthenium Homopolymer, NSC 71324Ruthenium(II) Chloride 1,5-Cyclooctadiene Complex Polymer
InChI Formula InChI=1S/C8H10.2ClH.Ru/c1-2-4-6-8-7-5-3-1;;;/h1,8H,2,4-5,7H2;2*1H;/q;;;+2/p-2
SMILES Cl[Ru]1(Cl)C2=CCCC1=CCC2
IUPAC Name 9,9-dichloro-9λ^{4}-ruthenabicyclo[3.3.1]nona-1,5-diene
Molecular Weight (g/mol) 278.14
Formula Weight 277.9203
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.