missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(chloromethyl)methylsilane, 95%, Thermo Scientific Chemicals

Product Code. 11905841
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1558-33-4
Molecular Formula C2H5Cl3Si
Molecular Weight (g/mol) 163.51
InChI Key JAYBZWYBCUJLNQ-UHFFFAOYSA-N
Synonym dichloro chloromethyl methylsilane, silane, dichloro chloromethyl methyl, ch3sicl2 ch2cl, chloromethyl methyldichlorosilane, chloromethylmethyldichlorosilane, unii-9p7n047n2s, chloromethyl dichloro methylsilane, dichloro chloromethyl methyl silane, silane cmm1, chloromethyldichloro methyl silane
PubChem CID 73788
IUPAC Name dichloro-(chloromethyl)-methylsilane
SMILES C[Si](CCl)(Cl)Cl

Specifications

Color Colorless to Straw
Density 1.2600g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2Si(Cl2)CH3
Refractive Index 1.4480 to 1.4510
Quantity 25 g
Specific Gravity 1.26
Solubility Information Solubility in water: reacts
Formula Weight 163.51
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dichloro(chloromethyl)methylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'