missing translation for 'onlineSavingsMsg'
Learn More

Dichloro-BPF, TRC

Product Code. 30380814
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380814 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380814 Supplier Toronto Research Chemicals Supplier No. TRCD438944100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloro-BPF
CAS 2056284-95-6
Molecular Formula C13H10Cl2O2
Quantity 100 mg
Synonym 2,6-Dichloro-4-(4-hydroxybenzyl)phenol, 2,6-Dichloro Bisphenol F
InChI Formula InChI=1S/C13H10Cl2O2/c14-11-6-9(7-12(15)13(11)17)5-8-1-3-10(16)4-2-8/h1-4,6-7,16-17H,5H2
SMILES Oc1ccc(Cc2cc(Cl)c(O)c(Cl)c2)cc1
Molecular Weight (g/mol) 269.12
Formula Weight 268.0058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.