missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II), 98%

Product Code. 11361469
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361469 10 g 10g
11371469 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361469 Supplier Thermo Scientific Alfa Aesar Supplier No. 013932.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14647-23-5 | C26H24Cl2NiP2 | 528.02 g/mol

Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II) is a reactant involved in Kumada catalyst-transfer polycondensation, oxidation of carboranyl phosphine ligands, synthesis of nickel-iron dithiolato hydrides, Ullmann reactions - homocoupling reactions, co-catalyst for hydroformylation of alcohols for the synthesis of quaternary carbon centers, catalyst for Wacker-type intramolecular aerobic oxidative amination of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14647-23-5
Molecular Formula C26H24Cl2NiP2
Molecular Weight (g/mol) 528.02
MDL Number MFCD00013313
InChI Key XXECWTBMGGXMKP-UHFFFAOYSA-L
Synonym 1,2-bis diphenylphosphino ethane nickel ii chloride
PubChem CID 131664338
IUPAC Name nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
SMILES Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 263°C to 265°C
Color Brown to Orange-Red
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 528.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.