missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer

Product Code. 11305998
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11305998

Brand: Thermo Scientific Alfa Aesar 010494.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50982-12-2 | C8H12Cl2Ru | 280.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50982-12-2
Molecular Formula C8H12Cl2Ru
Molecular Weight (g/mol) 280.15
MDL Number MFCD00171304
InChI Key DMRVBCXRFYZCPR-PHFPKPIQSA-L
Synonym dichloro cycloocta-1,5-diene ruthenium ii, dichloro 1,5-cyclooctadiene ruthenium ii, 1,5-cyclooctadiene, z,z-; dichlororuthenium
PubChem CID 91884784
IUPAC Name (5Z)-cycloocta-1,5-diene;dichlororuthenium
SMILES [Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1

Specifications

Melting Point 200°C (decomposition)
Color Brown to Black
Quantity 1 g
Physical Form Powder
Chemical Name or Material Dichloro(1,5-cyclooctadiene)ruthenium(II), Polymer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.