missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II), Pd 18.5%, Thermo Scientific™

Product Code. 11333477
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11333477

Brand: Thermo Scientific Alfa Aesar 018779.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19978-61-1
Molecular Formula C26H26Cl2P2Pd+2
Molecular Weight (g/mol) 577.762
MDL Number MFCD00015702
InChI Key LDJXFZUGZASGIW-UHFFFAOYSA-N
Synonym dichloro 1,2-bis diphenylphosphino ethane palladium ii
PubChem CID 131675886
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane;palladium(2+);dihydrochloride
SMILES C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Pd+2]

Specifications

Melting Point >360°C
Color Yellow
Linear Formula PdCl2[(C6H5)2PCH2CH2P(C6H5)2]
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 575.75
Concentration or Composition (by Analyte or Components) Pd 18.5%
Physical Form Powder
Chemical Name or Material Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.