missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane (1:1), Pd 13%, Thermo Scientific Chemicals

Product Code. 11359597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359597 1 g 1g
11339597 5 g 5g
11349597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11359597 Supplier Thermo Scientific Alfa Aesar Supplier No. 041225.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) is used for palladium-catalyzed coupling reactions. It serves as a catalyst and also employed in carbonylation reactions. Further, it is used in cross-coupling reaction and Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95464-05-4
Molecular Formula C35H30Cl4FeP2Pd
Molecular Weight (g/mol) 816.64
MDL Number MFCD00792899
IUPAC Name Bis(diphenylphosphino)ferrocene]palladium(II) chloride
SMILES [Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 275°C to 280°C
Quantity 1 g
Synonym Dichloro[1, 1′-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane complex
Solubility Information Insoluble in water.
Formula Weight 816.64
Physical Form Powder
Chemical Name or Material [1, 1′-Bis(diphenylphosphino)ferrocene]palladium(II) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.