missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals

Product Code. 11441440
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441440 1 g 1g
11451440 5 g 5g
11461440 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11441440 Supplier Thermo Scientific Alfa Aesar Supplier No. H26895.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

diphosphine 1,1'-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
diphosphine 1,1′-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67292-34-6
Molecular Formula C34H28Cl2FeNiP2
Molecular Weight (g/mol) 683.98
MDL Number MFCD00270284
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
IUPAC Name Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
SMILES [Fe].Cl[Ni]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 285°C
Quantity 1 g
Formula Weight 684
Percent Purity 99%
Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.