missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]cobalt(II), 98%, Thermo Scientific™

Product Code. 11359808
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359808 1 g 1g
11349808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11359808 Supplier Thermo Scientific Alfa Aesar Supplier No. 044274.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67292-36-8
Molecular Formula C34H28Cl2CoFeP2
Molecular Weight (g/mol) 684.22
MDL Number MFCD03427161
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride; CoCl2(dppf)
IUPAC Name [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride
SMILES [Fe].Cl[Co]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C
Color Green
Quantity 1 g
Formula Weight 684.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dichloro[1,1'-bis (diphenylphosphino)ferrocene]cobalt (II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.