missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II), Pd 14.1%, Thermo Scientific Chemicals

Product Code. 15407195
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
15407195 1 g 1g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15407195

Brand: Thermo Scientific Alfa Aesar 046383.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 917511-90-1
Melting Point 294°C to 300°C
Color Orange
Molecular Formula C34H52Cl2FeP2Pd
MDL Number MFCD11656081
Quantity 1 g
Synonym PdCl2(dcypf); [1,1'-Bis(dicyclohexylphosphino)ferrocene]dichloropalladium(II)
SMILES [Fe].Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)c1cccc1.C1CCC(CC1)P(C1CCCCC1)c1cccc1
IUPAC Name Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II)
Molecular Weight (g/mol) 755.90
Formula Weight 755.91
Concentration or Composition (by Analyte or Components) Pd 14.1%
Physical Form Crystalline Solid
Chemical Name or Material Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.