missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%, Thermo Scientific Chemicals

Product Code. 11356229
Change view
Click to view available options
Quantity:
0.1 g
1 g
Unit Size:
0.1g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356229 1 g 1g
11366229 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11356229 Supplier Thermo Scientific Alfa Aesar Supplier No. 044829.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Coupling reactions, Suzuki couplings

Coupling reactions, Suzuki couplings.Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene] palladium(II) is used as a catalyst in Suzuki-cross coupling reactions, which involve reaction of aromatic halides with methyliminodiacetic acid derivatives. it is also used as a catalyst for the preparation of chalcones. It is used in the preparation of (bisphosphinometallocene)-palladium(II) chloride complexes, which used as a starting material for the Buchwald-Hartwig reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95408-45-0
Molecular Formula C26H44Cl2FeP2Pd
Molecular Weight (g/mol) 651.75
MDL Number MFCD08064219
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym [1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride; PdCl2(dtbpf)
IUPAC Name Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
SMILES [Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C

Specifications

Color Red
Quantity 1 g
Solubility Information Soluble in dichloromethane and chloroform. Partially soluble in acetone,alcohols,tertrahydrofuran and dioxane.
Formula Weight 651.77
Concentration or Composition (by Analyte or Components) Pd 16.3%
Physical Form Crystalline
Chemical Name or Material Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.