missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicarbonylcyclopentadienylcobalt, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Dicarbonylcyclopentadienylcobalt Catalyst for the cyclotrimerization of alkynes. It mediates the [2+2+2]-cyclization of allenediynes to tricyclic systems. catalyzes the formation of pyridines from a mixture of alkynes and nitriles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 139°C to 140°C (710 mmHg) |
Flash Point | 26°C (80°F) |
Quantity | 1 g |
UN Number | UN3281 |
Sensitivity | Air sensitive |
Solubility Information | Insoluble in water. Soluble in common organic solvents. |
Formula Weight | 180.05 |
Physical Form | Liquid |
Chemical Name or Material | Dicarbonylcyclopentadienylcobalt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'