missing translation for 'onlineSavingsMsg'
Learn More

Dicarbonylcyclopentadienylcobalt

Product Code. 11352608
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352608 5 g 5g
11362608 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352608 Supplier Thermo Scientific Alfa Aesar Supplier No. 023136.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Catalyst for the cyclotrimerization of alkynes | CAS: 12078-25-0

Dicarbonylcyclopentadienylcobalt Catalyst for the cyclotrimerization of alkynes. It mediates the [2+2+2]-cyclization of allenediynes to tricyclic systems. catalyzes the formation of pyridines from a mixture of alkynes and nitriles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12078-25-0
MDL Number MFCD00009589

Specifications

Boiling Point 139°C to 140°C (710 mmHg)
Flash Point 26°C (80°F)
Quantity 5 g
UN Number UN3281
Sensitivity Air sensitive
Solubility Information Insoluble in water. Soluble in common organic solvents.
Formula Weight 180.05
Physical Form Liquid
Chemical Name or Material Dicarbonylcyclopentadienylcobalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.