missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Dibutyryl-cAMP (Sodium Salt)
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
Description
- Dibutyryl-cAMP is a cell-permeable cyclic AMP (cAMP) analog that activates cAMP-dependent protein kinases (Schwede et al.) This product is supplied as the sodium salt of the molecule
- Suppresses experimental autoimmune encephalomyelitis development by reducing demyelination and mobilizing neural stem cells in the subventricular zone towards the demyelinated plaques (Khezri et al.)
- Induces intrinsic axon growth in peripheral and central nervous systems, and morphological differentiation of astrocytes (Knott et al.; Imamura & Ozawa)
- Stimulates neurite outgrowth in PC12 cells (Maruoka et al.)
Specifications
Specifications
| CAS | 16980-89-5 |
| Synonym | Bucladesine; DC 2797 |
| Quantity | 250 mg |
| Molecular Formula | C18H23N5O8P · Na |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction