missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Dibutyryl-cAMP, sodium salt

Product Code. 30001207
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001207 10 mg 10mg
18034932 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001207 Supplier R&D Systems™ Supplier No. 1141/10
  • ONLINE EXCLUSIVE PRICE
    £132.63
    £117.60 / 10mg
    Save £15.03
    11% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cell-permeable cAMP analog

Cell-permeable analog of cAMP; activates cAMP-dependent protein kinases. Promotes differentiation of dopaminergic neurons (iDA) from hPSCs in combination with other reagents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16980-89-5
Molecular Formula C18H26N5NaO8P

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 491.37
Purity >98%
Molecular Weight (g/mol) 494.397
Chemical Name or Material Dibutyryl-cAMP, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.