missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyltin(IV) Oxide, TRC
Specifications
Specifications
| CAS | 818-08-6 |
| Molecular Formula | C8 H18 O Sn |
| Quantity | 1 g |
| Synonym | Dibutyltin oxide (6CI), Dibutyloxostannane, Axion CS 2455, Cotin 100, Di-n-butyltin oxide, Di-n-butyltin(II) oxide, Dibutyloxotin, Dibutylstannane oxide, Eurecat 9555, Eurecat 9560, FC 4201, Fascat 4201, Fomrez SUL 11A, Fomrez SUL 11B, Fomrez SUL 11C, MSCAT 01, MSCAT 02, NSC 28130, Neostann U 300, Stann BO, Tegokat 248, Tin dibutyl oxide, U 300, ZT 4201 |
| InChI Formula | InChI=1S/2C4H9.O.Sn/c2*1-3-4-2;;/h2*1,3-4H2,2H3;; |
| SMILES | CCCC[Sn](=O)CCCC |
| IUPAC Name | dibutyl(oxo)tin |
| Molecular Weight (g/mol) | 248.94 |
| Formula Weight | 250.038 |
| Chemical Name or Material | Dibutyltin-oxide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?