missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyltin Dilaurate, TRC
Specifications
Specifications
| Chemical Name or Material | Dibutyltin Dilaurate |
| CAS | 77-58-7 |
| Molecular Formula | C32 H64 O4 Sn |
| Quantity | 2.5 g |
| Synonym | Dibutyltin dilaurate (6CI), Stannane, dibutylbis(lauroyloxy)- (8CI), Stannane, dibutylbis[(1-oxododecyl)oxy]- (9CI), Tin, dibutylbis(lauroyloxy)- (7CI), Lauric acid, dibutylstannylene deriv. (8CI), A 201, ADK Stab BT 11, Addocat 201, Addocat 210, Advastab T 12P, BL 42A, BNT-CAT 425, BT 11, BT 18, BXX 3778, Baerostab DBTL/C, Butynorate, C 101, C 101 (stabilizer), CAT 11, CAT 11 (catalyst), CM-A, CP, CP (catalyst), Cata-Chek 820, Cotin 200, D 22, D 7103, D 80, D 80 (catalyst), DBTD, DBTDL, DBTL, DBTL-KS 1260, DC 176, DOD-D 82727, DXR 81, DY 12, DY 28, Dabco T 12, Dabco T 12N, Dabco T 12SL, Davainex, Desmorapid Z, Di-n-butyltin dilaurate, Dibutylbis(1-oxododecyloxy)stannane, Dibutylbis(laurato)tin, Dibutylbis(lauroyloxy)tin, Dibutylstannylene dilaurate, Dibutyltin didodecanoate, Dibutyltin n-dodecanoate, Dow Corning 176, Embilizer L 101, Fascat 4202, Foamate S 9, Fomrez SUL 4, Gleck TL, HG 0111, JF-NS-C 010, Jeffcat T 12, K 1 Type A, KS 120, KS 1260, KS 20, Keverkat DBTL 162, Kosmos 19, L 101, LL 101, Lankromark LT 173, Liocat 118, Liocat 119, Mark 1038, Mark BT 1, Mark BT 11, Mark BT 18, Mark DBTL, Meister ZL 4401, Metacure T 12, Metatin 712, Metatin K 712, NSC 2607, Neostann U 100, Niax T 12, Nitto 8501, Octaflow BT 71, Ongrostab BLTM, Reatinor 932, Reaxis C 218, SCAT 1, SCAT 1W, SD-L 101, SM 2014C, SUL 4, Stabilizer D 22, Stanclere DBTL, Stanclere TL, Stann BL, Stann SB 430A, Stavinor 1200SN, T 12, TK 218, TL 10, TL 10 (catalyst), TL 100, TL 1000, TL 1000 (catalyst), TMV 186, TN 12, TN 12 (catalyst), TVS Tin Lau, TVS-TL 700, TVS-TL 800, Tegokat 218, Thermolite T 12, Thorcat 401, Tin dibutyl dilaurate, TinStab BL 277, Tinostat, U 100, U 12, YL 101, YL 101 (catalyst), Yoke T 12, ZT 101, ZT 102, Dodecanoic acid, 1,1'-(dibutylstannylene) ester |
| InChI Formula | InChI=1S/2C12H24O2.2C4H9.Sn/c2*1-2-3-4-5-6-7-8-9-10-11-12(13)14;2*1-3-4-2;/h2*2-11H2,1H3,(H,13,14);2*1,3-4H2,2H3;/q;;;;+2/p-2 |
| SMILES | CCCCCCCCCCCC(=O)O[Sn](CCCC)(CCCC)OC(=O)CCCCCCCCCCC |
| IUPAC Name | [dibutyl(dodecanoyloxy)stannyl] dodecanoate |
| Molecular Weight (g/mol) | 631.5582 |
| Formula Weight | 632.3827 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?