missing translation for 'onlineSavingsMsg'
Learn More

Dibutylboron trifluoromethanesulfonate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific™

Product Code. 10525773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10525773 10 g 10g
10015792 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10525773 Supplier Thermo Scientific Acros Supplier No. 379000100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60669-69-4 , 60-29-7
Molecular Formula C9H18BF3O3S
Molecular Weight (g/mol) 274.11
MDL Number MFCD00009669
InChI Key FAVAVMFXAKZTMV-UHFFFAOYSA-N
Synonym dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane
PubChem CID 2724243
IUPAC Name dibutylboranyl trifluoromethanesulfonate
SMILES CCCCB(CCCC)OS(=O)(=O)C(F)(F)F

Specifications

Color Orange to Yellow
Density 0.8150g/mL
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CF3SO3B[(CH2)3CH3]2
Quantity 10 g
Fieser 07,91; 09,140; 16,109
Formula Weight 274.11
Percent Purity 98%
Physical Form Solution
Chemical Name or Material Dibutylboron trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Dibutylboron trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.