missing translation for 'onlineSavingsMsg'
Learn More

Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane

Product Code. 10297013 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10297013

Brand: Thermo Scientific Acros 378990250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60669-69-4 | C9H18BF3O3S | 274.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutylboron trifluoromethanesulfonate
CAS 60669-69-4, 75-09-2
Color Orange to Yellow
Density 1.2710g/mL
Flash Point 27°C
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C9H18BF3O3S
Linear Formula CF3SO3B[(CH2)3CH3]2
MDL Number MFCD00009669
Quantity 25 mL
Fieser 07,91; 09,140; 16,109
Specific Gravity 1.271
Synonym dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane
Solubility Information Solubility in water: reacts.
InChI Key FAVAVMFXAKZTMV-UHFFFAOYSA-N
SMILES CCCCB(CCCC)OS(=O)(=O)C(F)(F)F
IUPAC Name dibutylboranyl trifluoromethanesulfonate
Molecular Weight (g/mol) 274.11
PubChem CID 2724243
Formula Weight 274.11
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibutylboron trifluoromethanesulfonate in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.