missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl phosphite, 14.5-16% P

Product Code. 10337520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10337520 500 g Glass bottle 500g
10573701 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10337520 Supplier Thermo Scientific Acros Supplier No. 155065000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1809-19-4 | C8H19O3P | 194.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809-19-4
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.21
MDL Number MFCD00066633
InChI Key OSPSWZSRKYCQPF-UHFFFAOYSA-N
Synonym dibutyl phosphite, dibutyl phosphonate, phosphonic acid, dibutyl ester, di-n-butylphosphite, dibutoxyphosphine oxide, mobil dbhp, dibutyl hydrogen phosphonate, di-n-butyl hydrogen phosphite, dibutylfosfit, phosphorous acid, dibutyl ester
PubChem CID 6327349
IUPAC Name dibutoxy(oxo)phosphanium
SMILES CCCCO[P+](=O)OCCCC

Specifications

Density 0.9950g/mL
Boiling Point 118°C to 119°C (11.0 mmHg)
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula [CH3(CH2)3O]2P(O)H
Refractive Index 1.4225 to 1.4245
Quantity 500 g
Beilstein 01, I, 187
Specific Gravity 0.995
Solubility Information Solubility in water: slightly soluble
Formula Weight 194.21
Percent Purity 14.5 to 16% (P)
Physical Form Liquid
Chemical Name or Material Dibutyl phosphite, 96%
Show More Show Less
compliance-icons
Product Identifier
  • Dibutyl phosphite
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.