missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl phosphate, 97%

Product Code. 10678994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10678994 2.5 kg Glass bottle 2.5kg
10742294 250 g Glass bottle 250g
10626051 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10678994 Supplier Thermo Scientific Acros Supplier No. 406640025
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-66-4 | C8H19O4P | 210.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-66-4
Molecular Formula C8H19O4P
Molecular Weight (g/mol) 210.21
MDL Number MFCD00015245
InChI Key JYFHYPJRHGVZDY-UHFFFAOYSA-N
Synonym dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
PubChem CID 7881
IUPAC Name dibutyl hydrogen phosphate
SMILES CCCCOP(O)(=O)OCCCC

Specifications

Acidity 259 to 267mg KOH/g
Melting Point -13.0°C
Density 1.0580g/mL
Boiling Point 250.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3(CH2)3O)2P(O)OH
Refractive Index 1.4270 to 1.4290
Quantity 2.5 kg
Specific Gravity 1.058
Formula Weight 210.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dibutyl phosphate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dibutyl phosphate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.