missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl phosphate, 97%

Product Code. 10626051 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
This item is not returnable. View return policy

Product Code. 10626051

Brand: Thermo Scientific Acros 406645000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-66-4 | C8H19O4P | 210.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-66-4
Molecular Formula C8H19O4P
Molecular Weight (g/mol) 210.21
MDL Number MFCD00015245
InChI Key JYFHYPJRHGVZDY-UHFFFAOYSA-N
Synonym dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
PubChem CID 7881
IUPAC Name dibutyl hydrogen phosphate
SMILES CCCCOP(O)(=O)OCCCC

Specifications

Acidity 259 to 267mg KOH/g
Melting Point -13.0°C
Density 1.0580g/mL
Boiling Point 250.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3(CH2)3O)2P(O)OH
Refractive Index 1.4270 to 1.4290
Quantity 500 g
Specific Gravity 1.058
Solubility Information Solubility in water: 17.2g/L
Viscosity 75 mPa.s (20°C)
Formula Weight 210.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dibutyl phosphate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dibutyl phosphate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.