missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl L-Tartrate, TRC

Product Code. 30385611
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385611 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385611 Supplier Toronto Research Chemicals Supplier No. TRCD429540100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyl L-Tartrate
CAS 15763-01-6
Molecular Formula C12H22O6
Quantity 100 g
Synonym 2,3-Dihydroxy-(2R,3R)-butanedioic Acid 1,4-Dibutyl Ester, 2,3-Dihydroxy-[R-(R*,R*)]-butanedioic Acid Dibutyl Ester, Tartaric Acid Dibutyl Ester, (+)-Dibutyl Tartrate, (+)-Tartaric acid Dibutyl Ester, (2R,3R)-Di-n-butyl Tartrate, (2R,3R)-Dibutyl Tartrate, Di-n-butyl L-Tartrate, Dibutyl (+)-L-Tartrate, Dibutyl (R,R)-Tartrate, Dibutyl L-(+)-Tartrate, Dibutyl Tartrate, ENT 396, L-(+)-Tartaric Acid Dibutyl Ester
InChI Formula InChI=1S/C12H22O6/c1-3-5-7-17-11(15)9(13)10(14)12(16)18-8-6-4-2/h9-10,13-14H,3-8H2,1-2H3
SMILES CCCCOC(=O)[C@H](O)[C@@H](O)C(=O)OCCCC
Molecular Weight (g/mol) 262.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.