missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl ether, Reagent Grade, ≥99%, Honeywell™

Product Code. p-7111743 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
20 L
200 L
Packaging:
Can
Glass Bottle
Metal Drum
Unit Size:
1 L
20 L
200 L
This item is not returnable. View return policy

Product Code. 15671010

Brand: Honeywell Research Chemicals 1102801L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 142-96-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00009461
InChI Key DURPTKYDGMDSBL-UHFFFAOYSA-N
Synonym di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
PubChem CID 8909
IUPAC Name 1-butoxybutane
SMILES CCCCOCCCC

Specifications

CAS Max % ≤100.0000%
Melting Point -98°C (literature)
Density 0.764 g/mL at 25 °C(lit.)
Boiling Point 142°C to 143°C (literature)
Packaging Glass Bottle
Linear Formula [CH3(CH2)3]2O
Quantity 1 L
UN Number UN1149
Beilstein 1732752
Percent Purity >99%
Grade Reagent
Chemical Name or Material Dibutyl ether, Reagent Grade, ≥99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.