missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl ether, Reagent Grade, ≥99%, Honeywell™

Product Code. 15671010 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
20 L
200 L
Packaging:
Can
Glass Bottle
Metal Drum
Unit Size:
1 L
20 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15671010 1 L Glass Bottle 1 L
15601020 20 L Can 20 L
15691010 200 L Metal Drum 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15671010 Supplier Honeywell Research Chemicals Supplier No. 1102801L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 142-96-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00009461
InChI Key DURPTKYDGMDSBL-UHFFFAOYSA-N
Synonym di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
PubChem CID 8909
IUPAC Name 1-butoxybutane
SMILES CCCCOCCCC

Specifications

CAS Max % ≤100.0000%
Melting Point -98°C (literature)
Density 0.764 g/mL at 25 °C(lit.)
Boiling Point 142°C to 143°C (literature)
Packaging Glass Bottle
Linear Formula [CH3(CH2)3]2O
Quantity 1 L
UN Number UN1149
Beilstein 1732752
Percent Purity >99%
Grade Reagent
Chemical Name or Material Dibutyl ether, Reagent Grade, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.