missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Adipate, TRC

Product Code. 30383634
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383634 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383634 Supplier Toronto Research Chemicals Supplier No. TRCD4266105ML
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adipic acid-di-n-butyl ester
CAS 105-99-7
Molecular Formula C14 H26 O4
Quantity 5 mL
Synonym Adipic acid, dibutyl ester (6CI,7CI,8CI), Hexanedioic acid, dibutyl ester (9CI), 3PS, Adimoll DB, Adipic acid di-n-butyl ester, Butyl adipate, Cetiol B, Di-n-butyl adipate, Dibutyl adipate, Dibutyl hexanedioate, Experimental Tick Repellent 3, Experimental Tick Repellent 3PS, Monocizer W 260, NSC 8086, Polycizer W 260, W 260
InChI Formula InChI=1S/C14H26O4/c1-3-5-11-17-13(15)9-7-8-10-14(16)18-12-6-4-2/h3-12H2,1-2H3
SMILES CCCCOC(=O)CCCCC(=O)OCCCC
IUPAC Name dibutyl hexanedioate
Molecular Weight (g/mol) 258.35
Formula Weight 258.1831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.