missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl 3-Hydroxybutyl Phosphate, TRC

Product Code. 30384948
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384948 5 mg -
30387093 2.5 mg -
30376215 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384948 Supplier Toronto Research Chemicals Supplier No. TRCD4281555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89197-69-3
Molecular Formula C12 H27 O5 P
Molecular Weight (g/mol) 282.31
Synonym TBP-OH
IUPAC Name dibutyl 3-hydroxybutyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCC(C)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H27O5P/c1-4-6-9-15-18(14,16-10-7-5-2)17-11-8-12(3)13/h12-13H,4-11H2,1-3H3
Formula Weight 282.1596
Chemical Name or Material Dibutyl 3-Hydroxybutyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.