missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl 2,3,4,5-Tetrachlorophthalate, TRC

Product Code. 30381290
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381290 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381290 Supplier Toronto Research Chemicals Supplier No. TRCD4295105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyl 2,3,4,5-Tetrachlorophthalate
CAS 3015-66-5
Molecular Formula C16H18Cl4O4
Quantity 5 g
Synonym 3,4,5,6-Tetrachloro-1,2-benzenedicarboxylic Acid 1,2-Dibutyl Ester, 3,4,5,6-Tetrachloro-1,2-benzenedicarboxylic Acid Dibutyl Ester, Tetrachloro-phthalic Acid Dibutyl Ester, 3,4,5,6-Tetrachloro-1,2-benzenedicarboxylic Acid Dibutyl Ester, Dibutyl Tetrachlorophthalate
InChI Formula InChI=1S/C16H18Cl4O4/c1-3-5-7-23-15(21)9-10(16(22)24-8-6-4-2)12(18)14(20)13(19)11(9)17/h3-8H2,1-2H3
SMILES CCCCOC(=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1C(=O)OCCCC
IUPAC Name dibutyl 3,4,5,6-tetrachlorobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 416.12
Formula Weight 413.9959
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.