missing translation for 'onlineSavingsMsg'
Learn More

Dibucaine, MedChemExpress

Product Code. p-200100374
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16418528

Brand: Medchem Express HYB0552

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dibucaine (Cinchocaine) is a sodium channel inhibitor. Dibucaine is a potent SChE inhibitor.

Dibucaine (Cinchocaine) is a sodium channel inhibitor. Dibucaine is a potent SChE inhibitor.

Specifications

Chemical Name or Material Dibucaine
CAS 85-79-0
Color White
Purity Grade Notes Research
Molecular Formula C20H29N3O2
Quantity 10 g
Synonym Cinchocaine
Solubility Information DMSO : ≥ 36 mg/mL (104.82 mM)
SMILES O=C(C1=CC(OCCCC)=NC2=CC=CC=C12)NCCN(CC)CC
Molecular Weight (g/mol) 343.46
Formula Weight 343.46
Percent Purity 99.83%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.