missing translation for 'onlineSavingsMsg'
Learn More

Dibucaine Hydrochloride, TRC

Product Code. 30384564
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384564 100 mg -
30375774 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384564 Supplier Toronto Research Chemicals Supplier No. TRCD426500100MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-12-1
Molecular Formula C20 H29 N3 O2 . Cl H
Molecular Weight (g/mol) 379.92
Synonym 4-Quinolinecarboxamide, 2-butoxy-N-[2-(diethylamino)ethyl]-, monohydrochloride (9CI), Cinchoninamide, 2-butoxy-N-[2-(diethylamino)ethyl]-, monohydrochloride (8CI), 2-Butoxy-N-(2-diethylaminoethyl)cinchoninamide hydrochloride, 2-Butoxy-N-(2-diethylaminoethyl)cinchoninic acid amide hydrochloride, 2-n-Butoxy-N-(2-diethylaminoethyl)cinchoninamide hydrochloride, Benzolin, Benzolin (local anesthetic), Cincaine, Cincaine chloride, Cinchocaine hydrochloride, Cinchocainium chloride, Dibucaine hydrochloride, Nupercaine hydrochloride, Percain, Percaine, Percamin S, Sovcain, Sovcaine
IUPAC Name 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
SMILES Cl.CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H29N3O2.ClH/c1-4-7-14-25-19-15-17(16-10-8-9-11-18(16)22-19)20(24)21-12-13-23(5-2)6-3;/h8-11,15H,4-7,12-14H2,1-3H3,(H,21,24);1H
Formula Weight 379.2027
Chemical Name or Material Cinchocaine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.