missing translation for 'onlineSavingsMsg'
Learn More

Dibromocyanoacetamide, TRC

Product Code. 30383779
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383779 10 g
30388464 5 g
30382306 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383779 Supplier Toronto Research Chemicals Supplier No. TRCD42531010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10222-01-2
Molecular Formula C3 H2 Br2 N2 O
Molecular Weight (g/mol) 241.87
Synonym 2,2-Dibromo-2-cyanoacetamide, 2,2-Dibromo-3-nitrilopropanamide, 2,2-Dibromo-3-nitrilopropionamide, 2-Cyano-2,2-dibromoacetamide, AM 7287, Amerstat 300, BE 3S, Biomate 723, Busan 94, DB 20, DBNPA, DBNPA 7287, Dibromocyanoacetamide, Dibromonitrilopropionamide, Dow antimicrobial 7287, Dow bactericide 7287, Dowicil QK 20, Mucosin NT, NSC 98283, Odycide B 340, QK 20, Thor DB 20, X-CIDE 508, X-CIDE 509
IUPAC Name 2,2-dibromo-2-cyanoacetamide
SMILES NC(=O)C(Br)(Br)C#N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H2Br2N2O/c4-3(5,1-6)2(7)8/h(H2,7,8)
Formula Weight 239.8534
Chemical Name or Material Dibromocyanoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.