missing translation for 'onlineSavingsMsg'
Learn More

Dibromochloromethane, 97+%

Product Code. 11438776
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438776 100 g 100g
11418776 5 g 5g
11428776 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438776 Supplier Thermo Scientific Alfa Aesar Supplier No. A16938.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-48-1 | CHBr2Cl | 208.277 g/mol

It affords bromochlorocarbene under phase-transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It affords bromochlorocarbene under phase-transfer conditions.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-48-1
Molecular Formula CHBr2Cl
Molecular Weight (g/mol) 208.277
MDL Number MFCD00000820
InChI Key GATVIKZLVQHOMN-UHFFFAOYSA-N
Synonym chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform
PubChem CID 31296
ChEBI CHEBI:34627
IUPAC Name dibromo(chloro)methane
SMILES C(Cl)(Br)Br

Specifications

Melting Point -22°C
Density 2.42
Boiling Point 115°C to 118°C
Refractive Index 1.5475
Quantity 100 g
Beilstein 1731046
Sensitivity Light and moisture sensitive
Merck Index 14,2135
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 208.28
Percent Purity 98%
Chemical Name or Material Dibromochloromethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.