missing translation for 'onlineSavingsMsg'
Learn More

Dibromobis(triphenylphosphine)nickel(II), 99%, Thermo Scientific Chemicals

Product Code. 11399837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399837 25 g 25g
11309847 1 g 1g
11319847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399837 Supplier Thermo Scientific Alfa Aesar Supplier No. 042192.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a polymerization catalyst

As a polymerization catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14126-37-5
Molecular Formula C36H30Br2NiP2
Molecular Weight (g/mol) 743.09
MDL Number MFCD00015865
InChI Key QEKXARSPUFVXIX-UHFFFAOYSA-L
Synonym nickel ii bromide bis triphenylphosphine, nibr2 pph3 2, dibromobis triphenylphosphine nickel ii, bis triphenylphosphine nickel ii bromide, dibromobis triphenylphosphine nickel, dibromonickel; bis triphenylphosphine, bis triphenylphosphine dibromonickel ii
PubChem CID 11050914
IUPAC Name dibromonickel;triphenylphosphane
SMILES Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 219°C to 223°C
Odor Odorless
Quantity 25 g
Formula Weight 743.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.